Lanthanum standard solution, 1000 mg/L La in 5% nitric acid (from La₂O₃), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific

13870.AD 13870.AE 13870.AP
13870.ADEA 57.3 EUR
13870.AD 13870.AE 13870.AP
Lanthanum standard solution, 1000 mg/L La in 5% nitric acid (from La₂O₃), standard for ICP, Specpure®
Standards Cationic Standards Lanthanum Standards

Molecular Formula: La₂O₃ in 5% HNO₃ Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011066


Certifikace: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR