Titanium 99,99 Metals basis, rods, Ø 6.4 mm (0.25 in)

Supplier: Thermo Fisher Scientific
Warning

14001.BQ 14001.BU 14001.CZ
14001.BQEA 105 EUR
14001.BQ 14001.BU 14001.CZ
Titanium 99,99 Metals basis, rods, Ø 6.4 mm (0.25 in)
Titanium

Purity: 99.99% CAS No.: 7440-32-6 Physical Form: ≈1.45g/cm Odour: Odourless MDL No.: MFCD00011264

Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR