Laboratory and glassware washers, PG 8535 and PG 8536
Supplier: MIELE
7339400 7339410 7339390 7339380 7339370 7339360 7321240 7870410 7870440 9062950 7831480 7870450 7831710 9062920 7831680 7831500 7831700 7831490 7870430 7870460 7907620 9358490 7870470 7870420 9062900 7870490 7870510
141-0569EA 0 EUR
141-0569 141-0570 141-0568 141-0567 141-0566 141-0565 141-0560 141-0603 141-0606 141-0638 141-0590 141-0607 141-0597 141-0635 141-0595 141-0592 141-0596 141-0591 141-0605 141-0608 141-0612 141-0644 141-0609 141-0604 141-0633 141-0610 141-0611
Laboratory and glassware washers, PG 8535 and PG 8536
Glassware Washing Machines
These cleaning and disinfection machines with integrated drying system and drawer have been designed for the treatment of clinical instruments and for washing laboratory glassware in research laboratories, biology and pharmaceutical industries. The new PG 85 range offers a wide range of innovations such as freely programmable PROFITRONIC+ controls, PerfectSpeed spray arm monitoring, temperature independent PerfectFlow dispensing control on the basis of ultrasound as well as PerfectPureSensor conductivity metering.
  • Freely programmable PROFITRONIC+ controls with 64 program slots and
More Product Information
Order Now
SPECIFICATIONS
Model PG 8535 D PG 8535 D PG 8535 D PG 8536 D PG 8536 D PG 8536 D PG 8536 D PG 8536 D PG 8535 D PG 8536 D PG 8536 D PG 8535 D PG 8535 D PG 8536 D PG 8535 D PG 8536 D PG 8535 D PG 8536 D PG 8536 D PG 8535 D PG 8536 D PG 8535 D PG 8535 D PG 8536 D PG 8536 D PG 8535 D PG 8535 D
Environmentally Preferable
Capacity Example: 37 narrow-necked glassware items or 96 pipettes Example: 37 narrow-necked glassware items or 96 pipettes Example: 37 narrow-necked glassware items or 96 pipettes Example: 66 narrow-necked glassware items or 96 pipettes Example: 66 narrow-necked glassware items or 96 pipettes Example: 66 narrow-necked glassware items or 96 pipettes Example: 66 narrow-necked glassware items or 96 pipettes Example: 66 narrow-necked glassware items or 96 pipettes Example: 37 narrow-necked glassware items or 96 pipettes Example: 66 narrow-necked glassware items or 96 pipettes Example: 66 narrow-necked glassware items or 96 pipettes Example: 37 narrow-necked glassware items or 96 pipettes Example: 37 narrow-necked glassware items or 96 pipettes Example: 66 narrow-necked glassware items or 96 pipettes Example: 37 narrow-necked glassware items or 96 pipettes Example: 66 narrow-necked glassware items or 96 pipettes Example: 37 narrow-necked glassware items or 96 pipettes Example: 66 narrow-necked glassware items or 96 pipettes Example: 66 narrow-necked glassware items or 96 pipettes Example: 37 narrow-necked glassware items or 96 pipettes Example: 66 narrow-necked glassware items or 96 pipettes Example: 37 narrow-necked glassware items or 96 pipettes Example: 37 narrow-necked glassware items or 96 pipettes Example: 66 narrow-necked glassware items or 96 pipettes Example: 66 narrow-necked glassware items or 96 pipettes Example: 37 narrow-necked glassware items or 96 pipettes Example: 37 narrow-necked glassware items or 96 pipettes
Controller/programs Freely programmable Profitronic+ controller with 64 program slots and 16 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs Freely programmable Profitronic+ controller with 64 program slots and 16 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs Freely programmable Profitronic+ controller with 64 program slots and 16 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs Freely programmable Profitronic+ controller with 64 program slots and 17 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs Freely programmable Profitronic+ controller with 64 program slots and 17 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs Freely programmable Profitronic+ controller with 64 program slots and 17 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs Freely programmable Profitronic+ controller with 64 program slots and 17 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs Freely programmable Profitronic+ controller with 64 program slots and 17 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs Freely programmable Profitronic+ controller with 64 program slots and 16 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs Freely programmable Profitronic+ controller with 64 program slots and 17 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs Freely programmable Profitronic+ controller with 64 program slots and 17 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs Freely programmable Profitronic+ controller with 64 program slots and 16 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs Freely programmable Profitronic+ controller with 64 program slots and 16 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs Freely programmable Profitronic+ controller with 64 program slots and 17 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs Freely programmable Profitronic+ controller with 64 program slots and 16 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs Freely programmable Profitronic+ controller with 64 program slots and 17 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs Freely programmable Profitronic+ controller with 64 program slots and 16 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs Freely programmable Profitronic+ controller with 64 program slots and 17 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs Freely programmable Profitronic+ controller with 64 program slots and 17 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs Freely programmable Profitronic+ controller with 64 program slots and 16 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs Freely programmable Profitronic+ controller with 64 program slots and 17 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs Freely programmable Profitronic+ controller with 64 program slots and 16 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs Freely programmable Profitronic+ controller with 64 program slots and 16 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs Freely programmable Profitronic+ controller with 64 program slots and 17 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs Freely programmable Profitronic+ controller with 64 program slots and 17 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs Freely programmable Profitronic+ controller with 64 program slots and 16 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs Freely programmable Profitronic+ controller with 64 program slots and 16 standardised cleaning and disinfection programs; 39 vacant program slots for user-compiled programs
Program operation/display Touch screen control, clear display programs Touch screen control, clear display programs Touch screen control, clear display programs Touch screen control, clear display programs Touch screen control, clear display programs Touch screen control, clear display programs Touch screen control, clear display programs Touch screen control, clear display programs Touch screen control, clear display programs Touch screen control, clear display programs Touch screen control, clear display programs Touch screen control, clear display programs Touch screen control, clear display programs Touch screen control, clear display programs Touch screen control, clear display programs Touch screen control, clear display programs Touch screen control, clear display programs Touch screen control, clear display programs Touch screen control, clear display programs Touch screen control, clear display programs Touch screen control, clear display programs Touch screen control, clear display programs Touch screen control, clear display programs Touch screen control, clear display programs Touch screen control, clear display programs Touch screen control, clear display programs Touch screen control, clear display programs
Cleaning mechanisms Two spray arms (third spray arm on upper basket) Two spray arms (third spray arm on upper basket) Two spray arms (third spray arm on upper basket) Two spray arms (third spray arm on upper basket) Two spray arms (third spray arm on upper basket) Two spray arms (third spray arm on upper basket) Two spray arms (third spray arm on upper basket) Two spray arms (third spray arm on upper basket) Two spray arms (third spray arm on upper basket) Two spray arms (third spray arm on upper basket) Two spray arms (third spray arm on upper basket) Two spray arms (third spray arm on upper basket) Two spray arms (third spray arm on upper basket) Two spray arms (third spray arm on upper basket) Two spray arms (third spray arm on upper basket) Two spray arms (third spray arm on upper basket) Two spray arms (third spray arm on upper basket) Two spray arms (third spray arm on upper basket) Two spray arms (third spray arm on upper basket) Two spray arms (third spray arm on upper basket) Two spray arms (third spray arm on upper basket) Two spray arms (third spray arm on upper basket) Two spray arms (third spray arm on upper basket) Two spray arms (third spray arm on upper basket) Two spray arms (third spray arm on upper basket) Two spray arms (third spray arm on upper basket) Two spray arms (third spray arm on upper basket)
Metering devices 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 2× 5 l supply containers 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 2× 5 l supply containers 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 2× 5 l supply containers 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 4× 5 l supply containers 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 4× 5 l supply containers 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 4× 5 l supply containers 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 4× 5 l supply containers 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 4× 5 l supply containers 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 2× 5 l supply containers 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 4× 5 l supply containers 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 4× 5 l supply containers 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 2× 5 l supply containers 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 2× 5 l supply containers 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 4× 5 l supply containers 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 2× 5 l supply containers 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 4× 5 l supply containers 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 2× 5 l supply containers 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 4× 5 l supply containers 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 4× 5 l supply containers 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 2× 5 l supply containers 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 4× 5 l supply containers 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 2× 5 l supply containers 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 2× 5 l supply containers 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 4× 5 l supply containers 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 4× 5 l supply containers 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 2× 5 l supply containers 1 dispenser pump for liquid acidic agents, 1 dispenser pump for liquid detergent, ultrasound dispensing volume control, drawer for 2× 5 l supply containers
Circulation pump (Qmax) 400 L/min 400 L/min 400 L/min 600 ml/min 600 ml/min 600 ml/min 600 ml/min 600 ml/min 400 L/min 600 ml/min 600 ml/min 400 L/min 400 L/min 600 ml/min 400 L/min 600 ml/min 400 L/min 600 ml/min 600 ml/min 400 L/min 600 ml/min 400 L/min 400 L/min 600 ml/min 600 ml/min 400 L/min 400 L/min
Control Electric door lock, temperature control, Ao value control Electric door lock, temperature control, Ao value control Electric door lock, temperature control, Ao value control Electric door lock, temperature control, Ao value control Electric door lock, temperature control, Ao value control Electric door lock, temperature control, Ao value control Electric door lock, temperature control, Ao value control Electric door lock, temperature control, Ao value control Electric door lock, temperature control, Ao value control Electric door lock, temperature control, Ao value control Electric door lock, temperature control, Ao value control Electric door lock, temperature control, Ao value control Electric door lock, temperature control, Ao value control Electric door lock, temperature control, Ao value control Electric door lock, temperature control, Ao value control Electric door lock, temperature control, Ao value control Electric door lock, temperature control, Ao value control Electric door lock, temperature control, Ao value control Electric door lock, temperature control, Ao value control Electric door lock, temperature control, Ao value control Electric door lock, temperature control, Ao value control Electric door lock, temperature control, Ao value control Electric door lock, temperature control, Ao value control Electric door lock, temperature control, Ao value control Electric door lock, temperature control, Ao value control Electric door lock, temperature control, Ao value control Electric door lock, temperature control, Ao value control
Power connection Input voltage: AC 400 V, 50Hz - total electric connection: 9,7 kW Input voltage: AC 400 V, 50Hz - total electric connection: 9,7 kW Input voltage: AC 400 V, 50Hz - total electric connection: 9,7 kW Input voltage: AC 400 V, 50 Hz - total electric connection: 10,2 kW Input voltage: AC 400 V, 50 Hz - total electric connection: 10,2 kW Input voltage: AC 400 V, 50 Hz - total electric connection: 10,2 kW Input voltage: AC 400 V, 50 Hz - total electric connection: 10,2 kW Input voltage: AC 400 V, 50 Hz - total electric connection: 10,2 kW Input voltage: AC 400 V, 50Hz - total electric connection: 9,7 kW Input voltage: AC 400 V, 50 Hz - total electric connection: 10,2 kW Input voltage: AC 400 V, 50 Hz - total electric connection: 10,2 kW Input voltage: AC 400 V, 50Hz - total electric connection: 9,7 kW Input voltage: AC 400 V, 50Hz - total electric connection: 9,7 kW Input voltage: AC 400 V, 50 Hz - total electric connection: 10,2 kW Input voltage: AC 400 V, 50Hz - total electric connection: 9,7 kW Input voltage: AC 400 V, 50 Hz - total electric connection: 10,2 kW Input voltage: AC 400 V, 50Hz - total electric connection: 9,7 kW Input voltage: AC 400 V, 50 Hz - total electric connection: 10,2 kW Input voltage: AC 400 V, 50 Hz - total electric connection: 10,2 kW Input voltage: AC 400 V, 50Hz - total electric connection: 9,7 kW Input voltage: AC 400 V, 50 Hz - total electric connection: 10,2 kW Input voltage: AC 400 V, 50Hz - total electric connection: 9,7 kW Input voltage: AC 400 V, 50Hz - total electric connection: 9,7 kW Input voltage: AC 400 V, 50 Hz - total electric connection: 10,2 kW Input voltage: AC 400 V, 50 Hz - total electric connection: 10,2 kW Input voltage: AC 400 V, 50Hz - total electric connection: 9,7 kW Input voltage: AC 400 V, 50Hz - total electric connection: 9,7 kW
Water connection 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD) 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD) 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD) 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD) 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD) 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD) 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD) 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD) 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD) 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD) 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD) 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD) 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD) 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD) 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD) 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD) 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD) 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD) 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD) 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD) 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD) 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD) 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD) 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD) 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD) 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD) 1×cold water (EC), 1×cold water for vapour condenser, 1×hot water (AW), 1×demineralised water (AD)
W×D×H 900×700×820 mm 900×700×820 mm 900×700×820 mm 900×700×1175 mm 900×700×1175 mm 900×700×1175 mm 900×700×1175 mm 900×700×1175 mm 900×700×820 mm 900×700×1175 mm 900×700×1175 mm 900×700×820 mm 900×700×820 mm 900×700×1175 mm 900×700×820 mm 900×700×1175 mm 900×700×820 mm 900×700×1175 mm 900×700×1175 mm 900×700×820 mm 900×700×1175 mm 900×700×820 mm 900×700×820 mm 900×700×1175 mm 900×700×1175 mm 900×700×820 mm 900×700×820 mm
Chamber W×D×H 535×500×500 mm 535×500×500 mm 535×500×500 mm 535×500×500 mm 535×500×500 mm 535×500×500 mm 535×500×500 mm 535×500×500 mm 535×500×500 mm 535×500×500 mm 535×500×500 mm 535×500×500 mm 535×500×500 mm 535×500×500 mm 535×500×500 mm 535×500×500 mm 535×500×500 mm 535×500×500 mm 535×500×500 mm 535×500×500 mm 535×500×500 mm 535×500×500 mm 535×500×500 mm 535×500×500 mm 535×500×500 mm 535×500×500 mm
Weight 114 kg 114 kg 114 kg 177 kg 177 kg 177 kg 177 kg 177 kg 114 kg 177 kg 177 kg 114 kg 114 kg 177 kg 114 kg 177 kg 114 kg 177 kg 177 kg 114 kg 177 kg 114 kg 114 kg 177 kg 177 kg 114 kg 114 kg