Sporicide, Contec® ProChlor
Supplier: CONTEC
FBT102PC FBC502PC FBC102PC
141-0883EA 197 EUR
141-0883 141-0882 CONTFBC102PC
Sporicide, Contec® ProChlor
Cleaning Agents Disinfectants
Contec ProChlor is a fast acting innovative sporicidal biocide designed for use in pharmaceutical cleanrooms. A blend of hypochlorous acid in purified water. Ready-to-use format.
  • Efficacious against bacteria, fungi, moulds, yeasts, viruses and spores
  • Log 6 kill against Bacillus subtilis in 60 s saves time spent on biodecontamination
  • Filtered to 0,2 µm in a Grade B cleanroom ensures the product is free from contamination and particulates
  • Contains no quaternary ammonium or surfactant so it's very low residue
  • Trigger spray can be set to jet or spray - large