Mopping systems, VertiKlean®, MAX™
Supplier: CONTEC
VKMX0125 VKMX0225 VKMX0275 VKMX0175 VKMX0325 VKMX0375 VKSE0200 VKSE0250 VKSE0100 VKSE0150 VKSE0350 VKSE0300
141-0989EA 514 EUR
141-0989 141-0991 141-0992 141-0990 141-0993 141-0995 129-1477 129-1478 129-1195 129-1196 129-1480 129-1479
Mopping systems, VertiKlean®, MAX™
Mops
VertiKlean® MAX™ is designed for applying disinfectants to both horizontal and vertical surfaces and is ideal for general cleaning of ceilings, walls and floors. This lightweight mop features a single layer of hydrophilic polyurethane foam laminated to polyester microfibre fabric backed with a semi-rigid thermoformed plate, which easily snaps onto a VertiKlean® mop head frame.
  • Hydrophilic urethane foam and knitted polyester mop head retains liquids better than comparable products
  • Laser cutting seals the edges of the foam and polyester, reducing the chance