Lead titanate ≥99.5%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Lead titanium oxide, Lead(II) titanium oxide, Lead(II) titanate, Lead titanate (IV), Lead titanate

14139.22 14139.36 14139.A3
14139.22EA 54.4 EUR
14139.22 14139.36 14139.A3
Lead titanate ≥99.5%
Lead titanate

Purity: 99.5% CAS No.: 12060-00-3 Molecular Formula: PbTiO3 Formula Weight: 303.09 Physical Form: -325 Mesh Powder Density: 7.520 Appearance: Yellow Odour: Odourless UN No.: 3288 MDL No.: MFCD00011159

Formula: O₃PbTi
MW: 303.08 g/mol
Melting Pt: 1120
Density: 7.520
Storage Temperature: Ambient
MDL Number: MFCD00011159
CAS Number: 12060-00-3
EINECS: 235-038-9
ADR: 6.1 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR