Ultrasonic baths, Elmasonic P series
Supplier: ELMA SCHMIDBAUER
1013737 1013735 1013761 1013777 1013804 1013810
142-0051EA 1000 EUR
142-0051 142-0046 142-0047 142-0048 142-0049 142-0050
Ultrasonic baths, Elmasonic P series
Ultrasonic Baths
The Elmasonic P ultrasonic series is ideal for all applications in analytical and medical laboratories and in industrial production. Six different sizes are available. These practical units are easy to operate with a clearly arranged display showing all set and actual parameters. Several special features make the units perfect for intensive cleaning applications. All models are equipped with a mechanical timer up to maximum of 15 minutes.
  • Clear display
  • Easy and intuitive setting
  • Power-controlled ultrasound with high energy density for difficult