Centrifuge, benchtop, biomedical, LMC-3000
Supplier: GRANT INSTRUMENTS
LMC-3000
142-0125EA 1920 EUR
142-0125
Centrifuge, benchtop, biomedical, LMC-3000
Centrifuges Benchtop Centrifuges
The Grant LMC-3000 is a low speed benchtop centrifuge ideal for biomedical applications. It has a choice of interchangeable rotors for accommodating centrifuge tubes (10 to 15 ml or 50 ml) or standard 96-well microtitre plates.
  • Sturdy metal housing and lid; automatic imbalance switch-off and lid lock when centrifuge is running provide for safe operation
  • Easy set-up of spin speed and time via two line LCD display and simple push buttons
  • Timed operation (1 to 90 minutes) with automatic switch-off
  • Compact footprint that fits neatly into the