Manganese standard dissolved in oil, 1000 µg/g Mn in mineral oil for wear metal analysis, Specpure®

Supplier: Thermo Fisher Scientific

14214.18
14214.18EA 97.4 EUR
14214.18
Manganese standard dissolved in oil, 1000 µg/g Mn in mineral oil for wear metal analysis, Specpure®
Standards Cationic Standards Manganese Standards

Physical form: liquid
MDL No.: MFCD00011111

Order Now

SPECIFICATIONS

Environmentally Preferable
MDL No. MFCD00011111

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR