Beryllium standard dissolved in oil, 1000 µg/g Be in mineral oil for wear metal analysis, Specpure®

Supplier: Thermo Fisher Scientific

14221.1Y 14221.18
14221.1YEA 0 EUR
14221.1Y 14221.18
Beryllium standard dissolved in oil, 1000 µg/g Be in mineral oil for wear metal analysis, Specpure®
Standards Cationic Standards Beryllium Standards

Physical form: Liquid
MDL No.: MFCD00134032

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR