Manganese standard solution, 10000 mg/L Mn in 5% nitric acid (from Mn), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific

14413.AD 14413.AE 14413.AP
14413.ADEA 97.9 EUR
14413.AD 14413.AE 14413.AP
Manganese standard solution, 10000 mg/L Mn in 5% nitric acid (from Mn), standard for ICP, Specpure®
Standards Cationic Standards Manganese Standards

Molecular Formula: Mn in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011111


Certifikace: Traceable to NIST

Order Now

SPECIFICATIONS

Environmentally Preferable
MDL No. MFCD00011111

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR