Neutralising agent for the automated cleaning of glassware, neodisher® Z
Supplier: Dr. Weigert
420233 420230 420226 420287
148-3212EA 90.1 EUR
148-3212 148-3214 148-0164 WEIG420287
Neutralising agent for the automated cleaning of glassware, neodisher® Z
Cleaning Agents Detergents and Surface Cleansers
neodisher® Z
  • Acidic liquid concentrate for neutralisation of alkaline residues in the main automated cleaning cycle or for acidic pre-cleaning of laboratory glassware. Free of phosphates and surfactants
  • Components: Organic acids
  • Dosage: 1 to 2 ml/L for neutralisation; 2 to 5 ml/L at 40 to 60 °C for acidic pre-cleaning
Note: The materials are corrosive or irritant and care must be taken with the choice of materials for cleaning aluminium and titanium items. Please read the Material Safety Data sheet before use.