Ficoll® PM400
Supplier: Cytiva
17-0300-05 17-0300-08 17-0300-10 17-0300-50
17-0300-05EA 7184 EUR
17-0300-05 17-0300-08 17-0300-10 17-0300-50
Ficoll® PM400
Cell Separation Media
Ficoll® PM400 is a synthetic neutral, highly branched hydrophilic polymer of sucrose with an average molecular weight of 400000. It is used to form density gradients for separating and isolating eukaryotic cells, organelles and bacterial cells, as a stabilising agent and as a preparation medium for isolating mononuclear cells. It is useful for separating cells that are sensitive to centrifugation. Ficoll™ PM400 can also be used in other applications, such in defined culture media, nucleic acid hybridisation, electrophoresis, and immunological studies.
  • Concentrations of up to 50% (w/v)
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Stability Stable in alkaline and neutral solutions, but are rapidly hydrolyzed in solution at pH 3, especially at elevated temperature