Reversed phase FPLC columns, SOURCE™ 15RPC ST
Supplier: Cytiva
17-5068-01
17-5068-01EA 1170 EUR
17-5068-01
Reversed phase FPLC columns, SOURCE™ 15RPC ST
Chromatography Columns
Source 15RPC ST 4.6/100 is a prepacked stainless steel column which provides fast, high-capacity preparative protein purification with reversed phase chromatography.
  • Wide pH range (1 to 12), outstanding selectivity, chemical resistance, high capacity, and high resolution at high flow rates
  • Excellent scalability, from RESOURCE to FineLINE columns
  • Using ÄKTA design and other high-performance liquid chromatography systems
SOURCE 15RPC is based on rigid, monosized 15 µm diameter polystyrene/divinyl benzene beads. The matrix has outstanding selectivity forMore Product Information