Hydrophobic interaction chromatography media, Capto™ Phenyl (high sub)

Supplier: Cytiva

17-5451-01
17-5451-01EA 173 EUR
17-5451-01
Hydrophobic interaction chromatography media, Capto™ Phenyl (high sub)
Protein Purification Protein Purification Resins

Capto™ Phenyl (high sub) is used in the capture and intermediate stages of protein purification.


  • Improved productivity and process economy in downstream operations
  • Very high flow rates and large sample volume processing
  • Excellent chemical stability


The combination of high capacity, narrow specification range, high flow rate and low backpressure consistently reduces process cycle times and increases productivity.

Order Now

SPECIFICATIONS

Ligand Phenyl
Environmentally Preferable
Matrix Highly cross-linked agarose
pH range 3 - 13
Binding capacity 27 mg BSA/ml
Storage temperature 4 - 30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR