Water purification systems, UltraClear
Supplier: EVOQUA WATER TECHNOLOGIES
2001-D 2002-D 2001-D/60 2002-D/60 2002-D/80
171-0042EA 0 EUR
171-0042 171-0044 171-0166 171-0167 SGWA2002-D/80
Water purification systems, UltraClear
Water Purification Systems Pure and Ultra Pure Water Systems
This Ultra Clear™ TWF system produces reagent grade ultrapure water (Type I) directly from tap water. The pretreatment, RO module, deionisation cartridge, storage tank, UV oxidation chamber, polisher, ultrafilter, and sterile filter are all integrated into one convenient unit.
  • Flexible-hose dispenser lets users remotely dispense water from up to two feet away
  • Automatic flushing mechanism prevents contaminant build-up and extends service life
  • Unit can be either bench or wall mounted
  • Cartridge changes are quick and simple
  • Available with different tank sizes