Water purification systems, Super-Q® Plus
Supplier: Merck
CPMB02202 CPEX02201 ZFSQ24004 CDFC02203 ZFSQ24003 ZFSQ240P4
172-5057EA 985 EUR
172-5057 172-5058 MILLZFSQ24004 MILLCDFC02203 MILLZFSQ24003 MILLZFSQ240P4
Water purification systems, Super-Q® Plus
Water Purification Systems Pure and Ultra Pure Water Systems
Super-Q® Plus System is a high-flow water system designed to provide the 'final polish' to water which has been pretreated by reverse osmosis, distillation or deionization. The systems consistently produce ultrapure water quality which exceeds Type 1 ASTM/CAP/CLSI specifications.
  • Modular design for the production of ultrapure water
  • Hourly automatic recirculation of water to maintain water quality between uses
  • Super-Q® water is produced 'on demand', so there is no need to store water and risk degrading its quality over time
Typical applications include:More Product Information