VWR® Peristaltic Pump, Feeding
Supplier: VWR Collection
181-0590EA 2220 EUR
181-0590 181-0591 181-0592 181-0593
VWR® Peristaltic Pump, Feeding
Pumps Peristaltic Pumps
VWR® peristaltic pump variants offer a selection of feeding pumps with different pump heads integrated into the pump housing. They cover a broad spectrum of application areas in the laboratory and in industry.
  • Pump heads are made of durable PPS plastic, single - and multi-channel pump heads, are integrated into the pump housing
  • Precise feeding with right-left rotation due to the precisely adjustable motor
  • Operating with brushless DC motor, 110 - 240 VDC, 50/60 Hz
  • Robustness and stability prevent pulsation oscillations and vibrations
  • Control of the
More Product Information
Order Now
SPECIFICATIONS
Model PFP 5116 PFP 5124 PFP 5408 PFP 5808
Environmentally Preferable
Channels 1 1 4 8
Reversible + + + +
Min. flow rate 0 ml/min 0 ml/min 0 ml/min 0 ml/min
Max. flow rate 1,415 ml/min 4,200 ml/min 53 ml/min 53 ml/min
Speed range 0 - 400 min⁻¹ 0 - 400 min⁻¹ 0 - 100 min⁻¹ 0 - 100 min⁻¹
IP rating IP 31 IP 31 IP 31 IP 31
W×D×H 161×315×240 mm 161/320/240 mm 161×335×240 mm 161×380×240 mm
Weight 4,1 kg 4,7 kg 4,7 kg 5,4 kg