Tin(II) 2-ethylhexanoate, tech.

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Ethylcaproic acid tin (II) salt, Tin bis(2-ethylhexanoate), Stannous 2-ethylhexanoate, 2-Ethylhexanoic acid tin (II) salt

18590.22 18590.36 18590.A3 AB204036.0100
18590.22EA 50.4 EUR
18590.22 18590.36 18590.A3 ABCRAB204036.0100
Tin(II) 2-ethylhexanoate, tech.
Tin (II) 2-ethylhexanoate

CAS No.: 301-10-0 Molecular Formula: C16H30O4Sn Formula Weight: 405.11 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.251 Refractive Index: 1.4940 Appearance: Colourless to pale yellow MDL No.: MFCD00002676

Formula: C₁₆H₃₀O₄Sn
MW: 405.12 g/mol
Density: 1.251
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002676
CAS Number: 301-10-0
EINECS: 206-108-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR