Vacuum pump system SC 920 G
Supplier: KNF
SC920G
189-0033EA 5287 EUR
189-0033
Vacuum pump system SC 920 G
Pumps Vacuum Diaphragm Pumps
Vacuum Pump System SC 920 G is speed regulated.
Application: Rotary evaporation, distillation.
  • Automatic, accurate recognition and monitoring of the boiling point using the integrated ramp function
  • Speed-controlled
  • PPS pump head combined with PTFE-coated diaphragm are ideal for aggressive/corrosive gases and vapors
  • Integrated gas ballast valve
  • Quiet operation
The vacuum pump system consists of a vacuum pump with electronic control, a suction-side separator, a high performance condenser with separator on the pressure side, and a hand terminal for remoteMore Product Information