Clamps for conical standard ground joints, spring steel, DURAN®
Supplier: DWK Life Sciences
290300202 290300305 290300408 290300605 290300708 290300802 290300905 290301104
SCOT290300202EA 5.5 EUR
SCOT290300202 201-0385 SCOT290300408 201-0386 201-0387 201-0388 201-0389 201-0140
Clamps for conical standard ground joints, spring steel, DURAN®
Clamps NS Joint Clamps
Stainless steel. Clamps for tight and secure fit of conical ground joint connections.
  • Very good chemical resistance, corrosion resistant
  • Ideal for use at very high temperatures
  • Dimensionally stable, easy to assemble and dismantle