Air/steam inlet tubes, straight, Quickfit®
Supplier: SciLabware
MF15/1 MF15/2 MF15/3
201-3056EA 21.9 EUR
201-3056 201-3057 201-3058
Air/steam inlet tubes, straight, Quickfit®
Tubes Gas Inlet Tubes
Pyrex® borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Cone with stem and reduced shank
Useful for construction of gas inlet tubes.
Stem length: 150 mm
Three shank lengths: 45/60/70 mm.