Dropping funnels, cylindrical, without pressure balance, Quickfit®
Supplier: SciLabware
D1/11GP D1/21GP D1/22GP D1/32GP
201-8315EA 117 EUR
201-8315 201-8316 201-8317 201-8318
Dropping funnels, cylindrical, without pressure balance, Quickfit®
Funnels Dropping Funnels
PYREX®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With socket and stem with cone, with GP Rotaflo stopcock.