Phenol 99.0-100.5%, detached crystals Ph. Eur.

Supplier: VWR Chemicals
Danger

Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene

20596.297EA 183 EUR
20596.297 20596.468
Phenol 99.0-100.5%, detached crystals Ph. Eur.
Phenol
Formula: C₆H₅OH
MW: 94.11 g/mol
Boiling Pt: 181.8 °C (1013 hPa)
Melting Pt: 40.8 °C
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 81 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002143
CAS Number: 108-95-2
EINECS: 203-632-7
UN: 1671
ADR: 6.1 II
REACH: 01-2119471329-32
Merck Index: 12,07390

Order Now

Specification Test Results

Assay 99.0 - 100.5 %
Appearance Conforms (see CoA/CoS)
Identification A Passes test
Identification B Passes test
Identification C Passes test
Appearance of solution Passes test
Acidity Passes test
Freezing point Min. 39.5 °C
Residue on evaporation Max. 0.05 %
Residual solvents Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR