tert-Butanol 99,5, AnalaR NORMAPUR® Reag. Ph. Eur. zA

Supplier: VWR Chemicals
Danger

Synonyms: Trimethylcarbinol, tert-Butyl alcohol, 2-Methyl-2-propanol

20814.292EA 67.3 EUR
20814.292 20814.460
tert-Butanol 99,5, AnalaR NORMAPUR® Reag. Ph. Eur. zA
tert-Butanol
Formula: (CH₃)₃COH
MW: 74.12 g/mol
Boiling Pt: 82.2 °C (1013 hPa)
Melting Pt: 25.3 °C
Density: 0.7887 g/cm³ (20 °C)
Flash Pt: 11
Storage Temperature: Ambient
MDL Number: MFCD00004464
CAS Number: 75-65-0
EINECS: 200-889-7
UN: 1120
ADR: 3 II
REACH: 01-2119444321-51
Merck Index: 12,01577
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63a1b0c51eec643e5d5b7152a3e410db93d1626e&vid=999328a3c31895b190d09313b24501682921af01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.5 %
Acidity Max. 0.0002 meq/g
Boiling point 81.5 - 83.0 °C
Colouration Max. 10 APHA
Density (26/4) 0.770 - 0.780
Solidification point 24 - 26 °C
Non-volatile residue Max. 20 ppm
Water Max. 0.1 %
Conforms to Reag. Ph.Eur. Passes test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR