Isobutanol 99,0, AnalaR NORMAPUR® Reag. Ph. Eur., ACS zA
Supplier: VWR Chemicals
Danger
Synonyms: iso-Butyl alcohol, Isobutylalcohol, iso-Butanol, 2-Methyl-1-propanol
20833.297EA 86.2 EUR
20833.297 20833.550
Isobutanol 99,0, AnalaR NORMAPUR® Reag. Ph. Eur., ACS zA
Isobutanol
Formula: (CH₃)₂CHCH₂OH
MW: 74.12 g/mol
Boiling Pt: 107.9 °C (1013 hPa)
Melting Pt: –108 °C
Density: 0.802 g/cm³ (20 °C)
Flash Pt: 28
Storage Temperature: Ambient
MDL Number: MFCD00004740
CAS Number: 78-83-1
EINECS: 201-148-0
UN: 1212
ADR: 3 III
Merck Index: 13,05148
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de7c83a107c7a71fa54ed2b1bbdfb78617fe3f67&vid=df3ce90fc7ae8dafb7fabe31b244825735325030&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay Min. 99.0 %
Acidity Max. 0.0006 meq/g
Boiling point 107 - 109 °C
Colouration Max. 10 APHA
Density (20/4) 0.799 - 0.803
Density (20/20) 0.800 - 0.804
Refractive index (15°C) 1.397 - 1.399
Refractive index (20°C) 1.395 - 1.396
Evaporation residue Max. 50 ppm
Water Max. 0.5 %
Conforms to Reag. Ph.Eur. Passes test