Methanol 99,8, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient HPLC

Supplier: VWR Chemicals
Danger

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

Filter 0,2 um_Nitrogen_filled

20864.320EA 89.4 EUR
20864.320 20864.420 20864.290 20864.360 20864.400 20864.910
Methanol 99,8, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient HPLC
Methanol
Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.79 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 (6.1) II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (on Anhydrous Substance) Min. 99.8%
Acidity Max. 0.0003 meq/g
Alkalinity Max. 0.0002 meq/g
Boiling point 64 - 65 °C
Density (20/20) 0.791 - 0.793
Evaporation Residue Max. 5 ppm
Water Max. 0.03%
Absorbance (210 nm) Max. 0.7
Absorbance (220 nm) Max. 0.3
Absorbance (230 nm) Max. 0.15
Absorbance (235 nm) Max. 0.1
Absorbance (250 nm) Max. 0.02
Absorbance (260 nm) Max. 0.01
Transmittance (210 nm) Min. 20%
Transmittance (220 nm) Min. 50%
Transmittance (225 nm) Min. 68%
Transmittance (230 nm) Min. 74%
Transmittance (235 nm) Min. 80%
Transmittance (250 nm) Min. 95%
Transmittance (260 nm) Min. 98%
Transmittance (260 - 420 nm) Min. 98%
Fluorescence (as Quinine; 254 nm) Max. 1 ppb
Conforms Ph. Eur. R 1053200 Passes test
Conforms Ph. Eur. R1 1053201 Passes test
Conforms Ph. Eur. R2 1053202 Passes Test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR