Test tube racks
Supplier: NEOLAB
2-1630 2-1631 2-1632 2-1633 2-1634
211-2761EA 12.5 EUR
211-2761 211-2762 211-2760 211-2770 211-2763
Test tube racks
Racks Tube Racks
The rack features a fixed frame with slide rails and lock mechanism for the interchangeable tiers.
  • Easy handling, alphanumerically marked positions
  • Stackable and autoclavable up to 121 °C.
  • Two upper tiers are colour coded depending on the size of the openings