Strips of 8 PCR tubes and PCR caps
Supplier: Brand
781320 781340 781321 781322 781323 781324 781341 781342 781343 781344 781334 781325
211-3263EA 96.6 EUR
211-3263 211-3265 211-3270 211-3271 211-3272 211-3273 211-3274 211-3275 211-3276 211-3277 731-1250 731-1272
Strips of 8 PCR tubes and PCR caps
Tubes PCR Tubes
PP. Eight connected 0,2 ml tubes and domed or flat caps in strips of eight.
  • Flat caps are suitable for real-time PCR (qPCR)
  • Easy to open and close without tools
  • The tubes have a holding strap at one end, and the cover strips have a small lip on the side of each cap for careful, contamination-free opening
  • Free from DNase, RNase and DNA
Colour: Clear