Starch, soluble Potato, TECHNICAL

Supplier: VWR Chemicals

Synonyms: Starch, Vitex® iodine indicator, Amylodextrin

21152.234EA 67.8 EUR
21152.234 21152.291
Starch, soluble Potato, TECHNICAL
Starch, soluble
Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-84-9
EINECS: 232-686-4
Merck Index: 13,08878

Order Now

Specification Test Results

Water Max. 20.5 %
Fe (Iron) Max. 10 ppm
Pb (Lead) Max. 0.5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR