Centrifuge tubes
Supplier: NEUBERT VOLUME GLASSWAERE
0446-25-222100 0446-18-1618 0446-32-343100 0446-18-162100 0446-32-443100 0446-32-443150 0446-45-563145 0446-45-602135 0446-25-283100 0446-25-343100 0446-32-282100 0446-32-403115
212-0224EA 34.1 EUR
212-0224 525-0233 525-0236 NEUB0446-18-162100 NEUB0446-32-443100 NEUB0446-32-443150 NEUB0446-45-563145 NEUB0446-45-602135 NEUB0446-25-283100 NEUB0446-25-343100 NEUB0446-32-282100 NEUB0446-32-403115
Centrifuge tubes
Tubes Centrifuge Tubes
Heavy glass, round bottom, threaded neck.
Certifikace: Acc. to DIN 58970 (Part II)
Informace pro objednávání: Screw caps should to be ordered separately.