Flasks, pear shaped
Supplier: SciLabware
FP25/1 FP50/1 FP100/1 FP100/2 FP10/1 FP50/1/1A FP50/2 FP100/3 FP5/1
212-0599EA 23.9 EUR
212-0599 212-0601 212-0595 212-0596 212-0594 212-0602 212-0603 BIBBFP100/3 BIBBFP5/1
Flasks, pear shaped
Flasks Pear Shaped Flasks
PYREX®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for semi-micro distillations. With standard ground joint sleeve and white labelling field.