DURAN® Test Tubes with or without Rim
Supplier: DWK Life Sciences
261300302 261300602 261300808 261301307 261301607 261302106 261302303 261302603 261302809 261303308 261303608 261310106 261310303 261310603 261310809 261311102 261311308 261311608 261312604 261312801 261313309 261313609 261313806 261303805 261301101 261312107 261312304 261301204 261311205
212-1004EA 61.8 EUR
212-1004 212-1006 212-1008 212-1012 212-1015 212-1016 212-1018 212-1020 212-1022 212-1024 212-1026 212-1102 212-1104 212-1106 212-1108 212-1110 212-1112 212-1114 212-1120 212-1122 212-1124 212-1126 212-1128 212-1028 212-1010 212-1116 212-1118 212-0380 212-0381
DURAN® Test Tubes with or without Rim
Tubes Test Tubes
DURAN®, borosilicate glass 3.3, clear. Thick-walled, exceptional mechanical strength.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes