Ammonium hydrogen difluoride 98, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: Ammonium bifluoride, Ammonium hydrogen fluoride

21262.360EA 376 EUR
21262.360
Ammonium hydrogen difluoride 98, GPR RECTAPUR®
Ammonium hydrogen difluoride
Formula: NH₄HF₂
MW: 57.04 g/mol
Melting Pt: 127
Density: 0.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012018
CAS Number: 1341-49-7
EINECS: 215-676-4
UN: 1727
ADR: 8 II
Merck Index: 13,00498

Order Now

Specification Test Results

Assay Min. 98 %
Heavy metals (as Pb) Max. 0.03 %
Cl (Chloride) Max. 0.05 %
SiF6 (Hexafluorosilicate) Max. 0.3 %
Fe (Iron) Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR