Stretchable silicone lids for laboratory vessels, DURAN®
Supplier: DWK Life Sciences
291101119 291101127 291101135 291102115 291102123 291102131 291103111 291103128 291103136 291100011 291100028 291100036 291100044 291100052 291100069 291104116 291104124 291104132 291105112 291105129 291105137
213-0516EA 2.4 EUR
213-0516 213-0517 213-0518 213-0519 213-0520 213-0521 213-0522 213-0523 213-0524 213-0513 213-0514 213-0515 213-0525 213-0526 213-0527 213-0528 213-0529 213-0530 213-0531 213-0532 213-0533
Stretchable silicone lids for laboratory vessels, DURAN®
Closures Bottle Closures
These stretchable silicone lids are used to close round and square vessels and are available in three sizes and colours, as well as a set of three.
  • For covering and sealing laboratory vessels
  • Lids are made of stretchable, chemical and heat resistant silicone
  • Recommended usable temperature range is –40 to +180 °C
  • Due to excellent stretchability, DURAN® silicone lids can be used for a wide variety of containers of different diameters and shapes
  • Lids may be sterilised (steam/autoclave), and are lab washer compatible
  • Each lid features a marking area
  • Reusable