Erlenmeyer flasks, narrow or wide neck, with beaded rim, DURAN®
Supplier: DWK Life Sciences
212161407 212161707 212162403 212163202 212163605 212163905 212164404 212165409 212166302 212166808 212167307 212261405 212261705 212262401 212263209 212263603 212263903 212264402 212265407 212266309 219902702 212162806 212165306
214-1102EA 10.8 EUR
214-1102 214-1104 214-1106 214-1108 214-1110 214-1112 214-1114 214-1116 214-1118 214-1120 214-1122 214-1152 214-1154 214-1156 214-1158 214-1160 214-1162 214-1164 214-1166 214-1168 214-0337 214-1107 214-0338
Erlenmeyer flasks, narrow or wide neck, with beaded rim, DURAN®
Flasks Erlenmeyer Flasks
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Volume graduations and labelling area.
With printed Retrace Code (batch identification); 214-1102 without Retrace Code.
*Non DIN ISO size
DIN ISO 24450 wide mouth (except 214-1152 and 214-1168)
Certifikace: ISO 1773 narrow mouth
DIN ISO 24450 wide mouth