Flasks, round bottom, with standard ground joint
Supplier: VITLAB
107897 107797 107997
214-1422EA 113 EUR
214-1422 214-1421 214-1423
Flasks, round bottom, with standard ground joint
Flasks Round Bottom Flasks
PFA, translucent.
  • Very good chemical resistance
  • Excellent thermal resistance from −270 to +250 °C
  • Autoclavable
Can also be used as safety flasks for rotary evaporators.