Sterilin™ Containers, Bijou, with Screw Cap
Supplier: Sterilin
129B 129A 129BBAC M00525
215-0329EA 332 EUR
215-0329 215-0328 216-2250 SLINM00525
Sterilin™ Containers, Bijou, with Screw Cap
Sample Containers General Purpose Containers
Polystyrene, clear, with polyethylene screw cap. Store small-volume samples in Thermo Scientific™ Sterilin™ 7 ml Polystyrene Bijou Containers. Ideal for pediatric samples and other small-volume clinical applications. Aseptically manufactured. Not available in America.
  • Available pre-filled with boric acid (0,09 g) for the preservation of urine samples
  • Suitable for centrifugation at 7200×g
  • Aseptically manufactured
  • Available with glass coverslip, ideal for work on the culture of intracellular organisms (eg. Chlamydia trachomatis, Rickettsia etc)
  • In vitro use