Wash bottles, wide neck, with vent cap, Azlon®
Supplier: SciLabware
WGW234PVENT WGW235PVENT WGW236PVENT WGW237PVENT WGW238PVENT WGW239PVENT WGW443VT WGW444VT WGW445VT WGW446VT WGW447VT WGW448VT
215-3783EA 43.7 EUR
215-3783 215-3782 215-3778 215-3779 215-3780 215-3781 215-3784 215-3785 215-3786 215-3787 215-3788 215-3789
Wash bottles, wide neck, with vent cap, Azlon®
Bottles Wash Bottles
LDPE bottle, square shoulder, with PP vent cap.
  • DripLok® vapour venting valve prevents solvent drips
  • Closures feature precision jet-tip aperture for fine control
  • Colour coded for easy identification
  • Available in six colours
Bottles with wide mouth for easy filling. Fitted with colour coded closures that maintain their shape in use.