ANSI/SLAS microplate holders, DURAN®
Supplier: DWK Life Sciences
292330802 292331404
215-4446EA 271 EUR
215-4446 215-4447
ANSI/SLAS microplate holders, DURAN®
Racks Bottle Racks
A microplate sized holder that allow small DURAN® GL 25 bottles to be used with liquid handling robots.
  • Convenient – Microplate holders meet the Footprint Dimensions in the ANSI/SBS standards
  • Precise fit - Custom made for six 10 or 25 ml clear or amber DURAN GL 25 bottles in a 2×3 arrangement
  • High quality - Made of aluminum, milled for accuracy 'Made in Germany'
  • Flexible usage - Aluminum has high thermal conductivity, so can be used with microplate heating or cooling systems
  • Cost effective - Robust and reusable construction for a long service life
  • Future