Dropping bottles, glass, DURAN®
Supplier: DWK Life Sciences
232702404 232702461 232701708 232701765
215-6212EA 21 EUR
215-6212 215-6222 215-6211 215-6221
Dropping bottles, glass, DURAN®
Bottles Dropping Bottles
Soda-lime glass, clear or amber.
  • Good chemical and physical properties
  • For use in applications that do not involve high thermal stresses
  • Replaceable NS glass pipette with rubber suction bulb
Amber to protect light-sensitive substances.