Buckets, Round, PP
Supplier: SEMADENI
0701
216-1258EA 133 EUR
216-1258
Buckets, Round, PP
Buckets
Universal multipurpose bucket, made of PP, with spout, reinforced rim and handle.
  • Graduated, autoclavable
  • Very good thermal resistance
  • Very good chemical resistance