Tanks, rectangular, with cover, Nalgene®
Supplier: Thermo Fisher Scientific
14200-0002 14200-0005 14200-0010 14200-0015 14200-0020 14200-0045
216-1921EA 260 EUR
216-1921 216-1922 216-1923 216-1924 216-1925 NALG14200-0045
Tanks, rectangular, with cover, Nalgene®
Tanks Rectangular Tanks
These PP rectangular tanks with cover can significantly reduce evaporation and contamination.
  • Good stress-crack resistance
  • Suitable for use with many organic chemicals
  • Tanks can be modified with a factory-installed spigot
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification NAR Certificate of Retirement