Carboys, narrow neck, with screw cap and stopcock connection
Supplier: LAMAPLAST
1192 1191 1190 1187 1188 330
216-2136EA 102 EUR
216-2136 216-2135 216-2134 216-2131 216-2132 216-2133
Carboys, narrow neck, with screw cap and stopcock connection
Bottles Carboys
HDPE, translucent.
  • Very strong and durable
  • With handles
  • For food use
  • Stopcock connection ³/₄"
  • Stopcock must be ordered separately
Ideal for storing, transporting and decanting liquids.