Single wafer shipping boxes, for 150 mm wafer handling, H 22 series
Supplier: ENTEGRIS
H22-60-0615 H22-602-0615 H22-601-0615 H22-601-62C02 H22-60-62C02 H22-602-62C02
216-2357EA 0 EUR
216-2357 216-2360 216-2358 216-2359 216-2362 216-2361
Single wafer shipping boxes, for 150 mm wafer handling, H 22 series
Wafer Handling Supplies Wafer Shippers
For shipping and storage applications.
  • Holds one wafer face down in a conical pocket contacting only the edge of the wafer
  • Spring securely retains the wafer to minimise particle generation
  • Screw-on cover provides secure closure and easy opening
  • Large top surface provides a convenient space for labelling
  • Stackable shipping boxes allow efficient use of storage space