VWR® Serum/Sample Vials
Supplier: VWR Collection
216-3040EA 31.1 EUR
216-3040 216-3047 216-3048 216-3049 216-3050
VWR® Serum/Sample Vials
Vials
Crimp neck vials with flat bottom made from clear borosilicate glass.
  • Highly resistant against chemicals
  • High temperature resistance
  • Can be used as vaccine or injectable drug containers
  • Stoppers and caps have to be ordered separately
Ideal for long-term sample storage or lyophilisation.