Carboys, wide neck, LDPE, with screw cap and spigot, Nalgene®
Supplier: Thermo Fisher Scientific
2318-0010 2318-0020 2318-0050 2318-0065 2318-0130
216-4406EA 74 EUR
216-4406 216-4407 216-4408 216-4409 216-4410
Carboys, wide neck, LDPE, with screw cap and spigot, Nalgene®
Bottles Carboys
LDPE, translucent with white PP screw closure and PP spigot.
  • Good chemical resistance
  • Temperature resistant from −100 to +80 °C
  • Leakproof closure
Ideal for storing and dispensing distilled water, reagents and acids. Graduated. For better durability and leak resistance, the drain cock thread is moulded in one piece with the bottle, not welded. The 10, 20 and 25 litre sizes are equipped with convenient wide shoulder handles.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact
Environmental certification IRECS; ISO 14001 (Certificate Registration No. EMS 752603) NAR Certificate of Retirement