Cork stoppers assortment
Supplier: BRAEMSWIG
KS-SORTIMENT
217-9654EA 42.1 EUR
217-9654
Cork stoppers assortment
Stoppers Cork Stoppers
Natural cork, A1 quality
  • Lightweight, tough
  • Easy to drill holes into
  • Very economical
The assortment 217-9654 contains 10 stoppers each of Cat No. 217-1012/-1014/-1016/-1017/-1018/-1019/-1020/-1021/-1022 and 217-1024.