Barium sulphate 98, GPR RECTAPUR®
Supplier: VWR Chemicals
21766.292EA 90 EUR
21766.292 21766.463
Barium sulphate 98, GPR RECTAPUR®
Barium sulphate
Formula: BaSO₄
MW: 233.39 g/mol
Boiling Pt: 1600
Melting Pt: 1345 °C
Density: 4.5 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003455
CAS Number: 7727-43-7
EINECS: 231-784-4
Merck Index: 13,00997
Order Now
Specification Test Results
Assay Min. 98 %
Acidity Max. 0.002 meq/g
Heavy metals (as Pb) Max. 10 ppm
PO4 (Phosphate) Max. 10 ppm