Vydac 219TP Diphenyl, HPLC columns
Supplier: Avantor
219GCC510EA 542 EUR
219GCC510 219TP5210 219TP5215 219TP5415 219TP54 219TP510 219TP1010 219FSK1210 219GD52/N 219GK52/N
Vydac 219TP Diphenyl, HPLC columns
Chromatography Columns
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations. The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
  • Long column lifetime and negligible phase leaching
  • Reliable protein purifications, scalable from analytical to preparative scale
  • Referenced in a large number of patents and publications
Lowest capacity first generation diphenyl phase. Recommended for polypeptides with aromatic sideMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Carbon load 4%
Endcapped Yes
USP code L11